GC Analysis of Basic Drugs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after SPME using 85 μm Polyacrylate Fiber, Fast GC Analysis
GC Analysis of Basic Drugs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after SPME using 85 μm Polyacrylate Fiber, Fast GC Analysis
Conditions
sample/matrix 500 µg/L of each compound in 2 mL 0.05M phosphate buffer, pH=11, plus sodium chloride (NaCl) to 25% (w/v)
desorption process 250 °C for 3 min
extraction immersion, 450 rpm stirring at room temp. for 15 min
SPME fiber 85 µm Polyacrylate (57304)
column SLB-5ms, 20 m x 0.18 mm I.D. x 0.36 µm (28576-U)
oven 100 °C (1 min), 20 °C/min to 325 °C (5 min)
carrier gas helium, 1.5 mL/min. constant
liner 0.75 mm I.D. SPME
MSD interface 325 °C
Description
Categories Analytical Chromatography, SPME Applications, GC Applications, Drugs & Pharmaceuticals, Pharmaceuticals, Drugs of Abuse
Featured Industry Forensics and Toxicology
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC