GC Analysis of Bergamot Essential Oil on SLB-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Bergamot Essential Oil on SLB-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SLB-5ms, 10 m x 0.10 mm I.D., 0.10 µm (28465-U)
oven 50 °C, 14.0 °C/min to 250 °C
carrier gas hydrogen, 57.0 cm/sec, 184.2 kPa
sample bergamot oil, diluted 10:1 in hexane
injection 0.4 µL, 600:1 split
liner 0.75 mm ID
inj. temp. 300 °C
detector FID, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Other Notes Complementary products:
Syringes