GC Analysis of Blood Alcohols on a Packed Column (5% Carbowax® 20M on 60/80 Carbopack™ B)
GC Analysis of Blood Alcohols on a Packed Column (5% Carbowax® 20M on 60/80 °Carbopack™ B)
Conditions
column 6 ft.x 2 mm I.D. glass
packing 5% Carbowax 20M on 60/80 °Carbopack B
oven 85 °C
carrier gas helium, 20 mL/min
sample 0.05-0.10% each analyte in water
injection 1 µL
detector FID
Description
Categories Analytical Chromatography, GC Applications, Alcohols
Featured Industry Forensics and Toxicology
Legal Information Carbopack is a trademark of Sigma-Aldrich Co. LLC, Carbowax is a registered trademark of Union Carbide
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1-Propanol
  • Acetaldehyde
  • Acetone
  • Ethanol
  • Isopropanol
  • Methanol