Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Blood Alcohols on SUPELCOWAX® 10
GC Analysis of Blood Alcohols on SUPELCOWAX® 10
Conditions
column SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.50 µm (24284)
oven 35 C (1 min), 10 C/min to 125 C (1 min)
carrier gas helium, 1.0 ml/min constant
sample blood alcohols, each analyte at 0.08% in water
injection 0.5 µL, 100:1 split
liner 4 mm I.D. split, cup design
detector FID, 200 C
Description
Categories Analytical Chromatography, GC Applications, Drugs of Abuse, Alcohols
Featured Industry Forensics and Toxicology
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1-Propanol
  • 2-Butanol
  • 2-Propanol
  • Acetaldehyde
  • Acetone
  • Ethanol
  • Methanol