GC Analysis of BTEX, Undecane, and Tridecane on SLB®-IL100
GC Analysis of BTEX, Undecane, and Tridecane on SLB®-IL100
Conditions
column SLB-IL100, 30 m x 0.25 mm I.D., 0.20 µm (28884-U)
oven 110 °C
carrier gas helium, 26 cm/sec
sample neat mixture containing varying percentages of each component
injection 0.1 µL, 300:1 split
liner 4 mm I.D., split, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Petroleum
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Benzene
  • Ethyl benzene
  • m-Xylene
  • o-Xylene
  • p-Xylene
  • Toluene
  • Tridecane
  • Undecane