GC Analysis of C1-C12 Primary Alcohols on SLB®-IL111i, Isothermal Analysis
GC Analysis of C1-C12 Primary Alcohols on SLB®-IL111i, Isothermal Analysis
Conditions
column SLB-IL111i, 30 m x 0.25 mm I.D., 0.20 µm (29883-U)
oven 110 °C
carrier gas helium, 28 cm/sec
sample 12 component primary alcohols mix, each analyte at 500 µg/mL in methylene chloride
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 260
Description
Analysis Note The inert SLB IL111i ionic liquic column produces great peak shapes for many primary alcohols via an isothermal analysis. This is impressive for such a polar column.
Featured Industry Chemicals and Industrial Polymers
Food and Beverages
Petroleum
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC