GC Analysis of C18:0, C18:1, C18:2, and C18:3 FAME Isomers on SP™-2560
GC Analysis of C18:0, C18:1, C18:2, and C18:3 FAME Isomers on SP™-2560
Conditions
column SP-2560, 200 m x 0.25 mm I.D., 0.20 µm (29688-U)
oven (175 C)
carrier gas hydrogen
sample Mix of C18:0, C18:1 (from partially hydrogenated vegetable oil [PHVO]), C18:2, and C18:3 FAME isomers
injection 1.0 µL, 50:1 split
liner Split type, cup design
inj. temp. 250 C
detector FID, 250 C
Description
Analysis Note Partially hydrogenated vegetable oil, linoleic acid, and linolenic acid FAMEs analyzed on the 200 meter SP-2560 capillary GC column.
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC