GC Analysis of C18:0, C18:1, C18:2, and C18:3 FAME Isomers on SLB®-IL111
GC Analysis of C18:0, C18:1, C18:2, and C18:3 FAME Isomers on SLB®-IL111
Conditions
column SLB-IL111, 200 m x 0.25 mm I.D., 0.20 µm (29689-U)
oven 150 °C
carrier gas hydrogen
sample (Mix of C18:0, C18:1 (from partially hydrogenated vegetable oil [PHVO]), C18:2, and C18:3 FAME isomers)
injection 1µL, 50:1 split
liner Split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Partially hydrogenated vegetable oil, linoleic acid, and linolenic acid FAMEs analyzed on the 200 meter SLB-IL111 capillary GC column.
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC