GC Analysis of C18:1 FAMEs in Partially Hydrogenated Vegetable Oil (PHVO) on SLB®-IL111
GC Analysis of C18:1 FAMEs in Partially Hydrogenated Vegetable Oil (PHVO) on SLB®-IL111
Conditions
column SLB-IL111, 100 m x 0.25 mm I.D., 0.20 µm (29647-U)
oven 168 C isothermal
carrier gas hydrogen, 1 ml/min
sample partially hydrogenated vegetable oil FAMEs
injection 1 µL, 100:1 split
liner 4 mm I.D., split liner with cup (2051001)
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes