GC Analysis of C2-C7 Free Fatty Acids on Nukol™
GC Analysis of C2-C7 Free Fatty Acids on Nukol™
Conditions
column Nukol, 30 m x 0.25 mm I.D., 0.25 µm (24107)
oven 185 °C
carrier gas helium, 20 cm/sec @ 185 °C
sample Volatile Acid Standard Mix (46975-U), each analyte at 10 mM in deionized water
injection 1 µL, 100:1 split
inj. temp. 220 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acids, Organic Acids, Acids
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information Nukol is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Acetic acid
  • Butyric acid
  • Caproic acid
  • Heptanoic acid
  • Isobutyric acid
  • Isocaproic acid
  • Isovaleric acid
  • Propionic acid
  • Valeric acid