GC Analysis of C3-C5 Alcohols on Supel-Q™ PLOT
GC Analysis of C3-C5 Alcohols on Supel-Q™ PLOT
Conditions
column Supel-Q PLOT, 30 m x 0.53 mm I.D. (25462)
oven 35 °C (3 min), 16 C/min to 250 °C
carrier gas helium, 3 mL/min @ 35 °C
sample 5-Component C3-C5 alcohol standard
injection 0.1 µL
inj. temp. 200 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Alcohols
Featured Industry Chemicals and Industrial Polymers
Legal Information Supel-Q is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1-Butanol
  • 1-Pentanol
  • 1-Propanol
  • 2-Ethyl-1-butanol
  • 2-Methyl-1-propanol
  • 4-Methyl-2-pentanol