GC Analysis of C5-C15 n-Alkanes and BTEX on SLB®-IL111
GC Analysis of C5-C15 n-Alkanes and BTEX on SLB®-IL111
Conditions
column SLB-IL111, 30 m x 0.25 mm I.D., 0.20 µm (28927-U)
oven 65 C
carrier gas helium, 30 cm/sec
sample neat mixture of C5-C15 n-alkanes + BTEX, equal volumes
injection wet needle, 200:1 split
liner 4 mm I.D. FocusLiner(TM) inlet liner with taper
inj. temp. 250 C
detector FID, 265 C
Description
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Petroleum
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes