Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of cis C18:1 FAMEs in Partially Hydrogenated Vegetable Oil (PHVO) on SLB®-IL111 after HPLC Fractionation using Silver Ion
GC Analysis of cis C18:1 FAMEs in Partially Hydrogenated Vegetable Oil (PHVO) on SLB®-IL111 after HPLC Fractionation using Silver Ion
Conditions
column SLB-IL111, 100 m x 0.25 mm I.D., 0.20 µm (29647-U)
oven 168 C isothermal
carrier gas hydrogen, 1 ml/min
sample cis fraction of partially hydrogenated vegetable oil FAMEs, prepared using Ag-ion fractionation
injection 1 µL, 100:1 split
liner 4 mm I.D., split liner with cup (2051001)
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes