GC Analysis of cis-Chlordane Enantiomers on Astec® CHIRALDEX® G-BP
GC Analysis of cis-Chlordane Enantiomers on Astec® CHIRALDEX® G-BP
Conditions
column Astec CHIRALDEX G-BP, 10 m x 0.25 mm I.D., 0.12 µm (75031AST)
oven 170 C
carrier gas helium, 15 psi
sample 5 mg/mL in ethanolpeaks 1 & 2: cis-chlordane enantiomers
injection 0.2 µL, 100:1 split
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Pesticides
Featured Industry Agriculture
Chemicals and Industrial Polymers
Environmental
Food and Beverages
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes