GC Analysis of cis/trans C18:1, C18:2, and C18:3 FAME Isomers on SP™-2560
GC Analysis of cis/trans C18:1, C18:2, and C18:3 FAME Isomers on SP™-2560
Conditions
column SP-2560, 100 m x 0.25 mm I.D., 0.20 µm (24056) (24056)
oven 175 °C
carrier gas helium, 20 cm/sec @ 175 °C
sample Mixture of C18:1, C18:2, and C18:3 FAMEs in methylene chloride
injection 1.0 µL, 100:1 split
inj. temp. 210 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC