GC Analysis of cis/trans C18:1, C18:2, and C18:3 FAME Isomers on SP™-2560 (75 m x 0.18 mm I.D., 0.14 μm), Fast GC Analysis
GC Analysis of cis/trans C18:1, C18:2, and C18:3 FAME Isomers on SP™-2560 (75 m x 0.18 mm I.D., 0.14 μm), Fast GC Analysis
Conditions
column SP-2560, 75 m x 0.18 mm I.D., 0.14 µm (23348-U)
oven 180 °C, isothermal
carrier gas hydrogen, 25 cm/sec @ 180 °C
sample mixture of C18:1, C18:2, and C18:3 FAMEs in methylene chloride
injection 0.5 µL, split 100:1
liner 4 mm I.D. split, cup design
inj. temp. 220 °C
detector FID, 220 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes