GC Analysis of Class 3 Residual Solvents on Equity®-5
GC Analysis of Class 3 Residual Solvents on Equity®-5
Conditions
column Equity-5, 30 m x 0.53 mm I.D., 5.0 µm (28279-U) with 5 m intermediate polarity guard (25339)
oven 35 °C (15 min), 5 °C/min to 200 °C
carrier gas helium, 30 cm/sec @ 35 °C
sample 27 analytes, each at 3000 µg/mL in methanol
injection 1 µL33:1 split
liner single taper
inj. temp. 225 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Solvents
Featured Industry Pharmaceutical (small molecule)
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1-Butanol
  • 1-Propanol
  • 2-Butanone
  • 2-Propanol
  • 4-Methyl-2-pentanone
  • Acetic acid
  • Acetone
  • Anisole
  • Butyl acetate
  • Cumene
  • Diethyl ether
  • Dimethyl sulfoxide
  • Ethanol
  • Ethyl acetate
  • Ethyl formate
  • Heptane
  • Isoamyl alcohol
  • Isobutanol
  • Isobutyl acetate
  • Isopropyl acetate
  • Methyl acetate
  • Methyl-t-butyl ether
  • n-Amyl alcohol
  • Pentane
  • Propyl acetate
  • sec-Butanol
  • Tetrahydrofuran (THF)