GC Analysis of Closely Eluting Analytes from US EPA Method 8270 on SLB-5ms (30 m x 0.53 mm I.D., 0.50 μm)
GC Analysis of Closely Eluting Analytes from US EPA Method 8270 on SLB-5ms (30 m x 0.53 mm I.D., 0.50 μm)
Conditions
column SLB-5ms, 30 m x 0.53 mm I.D., 0.50 µm (28541-U)
oven 40 °C (2 min), 22 °C/min to 240 °C, 10 °C/min to 330 °C (1 min)
carrier gas helium, 30 cm/sec @ 200 °C, set using methane
sample 50 ng on-column of a 72 °Component semivolatile standard and 8 surrogate compounds, plus 6 internal standards (at 40 ng on-column), in methylene chloride
injection 0.5 µL, splitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector FID, 330 °C
Description
Categories Analytical Chromatography, GC Applications, Semivolatiles
Featured Industry Environmental
Other Notes Complementary products:
Syringes