GC Analysis of Cold Pressed Lemon Essential Oil on Equity®-5 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Cold Pressed Lemon Essential Oil on Equity®-5 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Equity-5, 15 m x 0.10 mm I.D., 0.10 µm (28083-U)
oven 75 C (1 min), 35 C/min to 200 C (1 min)
carrier gas helium, 45 cm/sec constant
sample Cold pressed lemon oil, neat
injection 0.10 muL300:1 split
liner 2 mm I.D., straight
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 3-Carene
  • 6-Umonene
  • Camphene
  • cis-3,7-Dimethyl-2,6-octadien-1-ol
  • Citronellal
  • Citronellyl acetate
  • Decanal
  • Dodecanal
  • Geranial
  • Geraniol
  • Geranyl acetate
  • Heptanal
  • Lauric aldehyde
  • Linalool
  • Myrcene
  • Neral
  • Nerol
  • Neryl acetate
  • Nonanal
  • Nonyl acetate
  • Octanal
  • Octanol
  • Oecanal
  • p-Cymene
  • Sabinene
  • Terpinen-4-ol
  • Terpinolene
  • trans-α-Bergamotene
  • y-Terpinene
  • α-Humulene
  • α-Phellandrene
  • α-Pinene
  • α-Terpinene
  • α-Terpineol
  • α-Thujene
  • β-Bisobolene
  • β-Caryophyllene
  • β-Pinene
  • γ-Terpinene
  • δ-Limonene