GC Analysis of Cold-Pressed Mandarin Essential Oil on SLB®-5ms
GC Analysis of Cold-Pressed Mandarin Essential Oil on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50C at 3C/min to 250C, held 10 min, at 10C/min to 270C, held 3 min
carrier gas He at 30.1 cm/s and pressure of 99.8 kPa
detector FID, 280 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (E)-ß-Ocimene
  • (E,E)-α-Farnesene
  • (Z,E)-Farnesol
  • Camphene
  • Decanal
  • Limonene
  • Linalol
  • Methyl N-methyl anthranilate
  • Myrcene
  • Octanal
  • Octanol
  • p-Cymene
  • Perilla aldehyde
  • Sabinene
  • ß-Caryophyllene
  • ß-Pinene
  • Terpinen-4-ol
  • Terpinolene
  • Thymol
  • trans-Sabinene hydrate
  • y-Terpinene
  • α-Phellandrene
  • α-Pinene
  • α-Sinensal
  • α-Terpinene
  • α-Terpineol
  • α-Thujene