GC Analysis of Column Bleed on SLB®-IL60 using FID, Comparison to 5 Polyethylene Glycol (PEG) Columns
GC Analysis of Column Bleed on SLB®-IL60 using FID, Comparison to 5 Polyethylene Glycol (PEG) Columns
Conditions
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
column 30 m x 0.25 mm I.D., 0.25 µm
oven 50 °C (2 min), 15 °C/min to column programmed temperature limit (10
carrier gas helium, 1 mL/min
sample methylene chloride
injection 1.0 µLsplitless
liner 4 mm I.D., split/splitless type, single taper wool packed FocusLiner™ design
inj. temp. 250 °C
detector FID, at column programmed temperature limit
Description
Featured Industry General Analytical
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC