GC Analysis of Column Bleed on SLB®-IL60 during Thermal Stress Test, Comparison to a Polyethylene Glycol (PEG) Column
GC Analysis of Column Bleed on SLB®-IL60 during Thermal Stress Test, Comparison to a Polyethylene Glycol (PEG) Column
Conditions
column PEG, 30 m x 0.25 mm I.D., 0.25 µm
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
oven 50 C (2 min), 15 C/min to 300 C (20 min)
carrier gas helium, 1 ml/min
sample methylene chloride
injection 1 muLsplitless
liner 4 mm I.D., split/splitless type, single taper wool packed FocusLiner(TM) design
inj. temp. 250 C
detector FID, 300 C
Description
Featured Industry General Analytical
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC