Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Distilled Lime Essential Oil on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Distilled Lime Essential Oil on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Equity-1, 15 m x 0.10 mm I.D., 0.10 µm (28039-U)
oven 75 C (1 min), 35 C/min to 200 C (1 min)
carrier gas helium, 45 cm/sec constant
sample distilled lime oil, neat
injection 0.10 µL, 300:1 split
liner 2 mm I.D., straight
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (-)-trans-Caryophyllene
  • (-)-β-Pinene
  • (+)-Terpinen-4-ol
  • (1R)-endo-(+)-Fenchyl alcohol
  • 1,4-Cineole
  • 3,7-Dimethyl-2,6-octadienyl acetate
  • Borneol
  • Camphene
  • Decanal
  • Geranial
  • Lauric aldehyde
  • Linalool
  • Myrcene
  • Neral
  • Neryl acetate
  • p-Cymene
  • Terpinen-1-ol
  • Terpinolene
  • trans-α-Bergamotene
  • α-Farnesene
  • α-Phellandrene
  • α-Pinene
  • α-Terpinene
  • α-Terpineol
  • β-Bisabolene
  • β-Terpineol
  • γ-Terpinene
  • γ-Terpineol
  • δ-Limonene