GC Analysis of Durability Test of Molded Thermogreen® LB-2 Septum on SLB-5ms
GC Analysis of Durability Test of Molded Thermogreen® LB-2 Septum on SLB-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 115 °C (15 min)
carrier gas helium, 30 cm/sec. @ 115 °C
sample Nonpolar Column Test Mix, each analyte at 500 µg/mL in methylene chloride (47300-U)
injection 1 µL, 100:1 split
liner 4 mm I.D., split, cup design, unpacked
inj. temp. 250 °C
detector FID, 360 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry General Analytical
Legal Information Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes