GC Analysis of Enantiomers on γ-DEX™ 120
GC Analysis of Enantiomers on γ-DEX™ 120
Conditions
column gamma-DEX 120, 30 m x 0.25 mm I.D., 0.25 µm (24307)
oven 90 °C, 1 °C/min to 135 °C, 5 °C/min to 240 °C
carrier gas helium, 20 cm/sec
sample 500 µg/mL in methylene chloride
injection 1 µL100:1 split
inj. temp. 300 °C
detector FID, 300 °C
Description
Categories Analytical Chromatography, Chiral Applications, GC Applications, Clinical, Forensic & Toxicology, Enantiomers, Herbicides, Alcohols, Drugs of Abuse
Featured Industry Agriculture
Chemicals and Industrial Polymers
Forensics and Toxicology
Legal Information DEX is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (±)-1-(2',4',6'-Trimethylphenyl)ethanol
  • 2-Ethylhexanoic acid
  • Methamphetamine
  • α-BHC