GC Analysis of Estrogenic Compounds on SLB®-5ms
GC Analysis of Estrogenic Compounds on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 175 °C (0.5 min), 15 °C/min to 300 °C (5 min)
carrier gas helium, 1 mL/min, constant
sample Estrogenic compounds, as methylated derivatives, 10 ppm in ethyl acetate
injection 0.5 µL, pulsed splitless (30 psi until 0.5 min, splitter open at 1.5 min)
liner 2 mm I.D., straight
inj. temp. 250 °C
MSD interface 330 °C
scan range m/z = 45-525
Description
Categories Analytical Chromatography, GC Applications, Hormones
Featured Industry Clinical
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Estriol
  • Estrone
  • α-Estradiol
  • β-Estradiol