GC Analysis of Explosives on SLB®-5ms (12 m x 0.18 mm I.D., 0.30 μm), Fast GC Analysis
GC Analysis of Explosives on SLB®-5ms (12 m x 0.18 mm I.D., 0.30 μm), Fast GC Analysis
Conditions
column SLB-5ms, 12 m x 0.18 mm I.D., 0.30 µm (28566-U)
oven 50 C (1 min), 15 C/min to 250 C
carrier gas helium, 0.8 ml/min, constant
sample explosives, 1 µg/mL in ethyl acetate
injection 0.5 µl pulsed splitless (30 psi until 0.2 min, splitter open at 0.75 min)
liner 2 mm ID focus liner w/taper
inj. temp. 220 C
detector MS
MSD interface 300 C
Description
Analysis Note The SLB-5ms column was found to give good peak shape and response for a variety of explosives. This applications shows the GC-MS analysis of 13 different explosives on a short, thick film column. This dimension was chosen to reduce analysis time while still providing good peak shape for early eluting compounds and good response for the trinitro compounds. A pulsed injection in combination with a narrow bore focus liner were used to increase the speed of sample transfer onto the column.
Categories Analytical Chromatography, GC Applications, Explosives
Featured Industry Chemicals and Industrial Polymers
Environmental
Forensics and Toxicology
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes