GC Analysis of FAMEs on SP™-2330
GC Analysis of FAMEs on SP™-2330
Conditions
column SP-2330, 30 m x 0.25 mm I.D., 0.20 µm (24019)
oven 200 °C
carrier gas helium, 20 cm/sec @ 200 °C
sample FAMEs from a natural source dissolved in chloroform
injection 0.5 µL, split 100:1
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • C14:0
  • C16:0
  • C16:1
  • C18:1
  • C18:2
  • C18:3
  • C20:1
  • C20:3
  • C20:4
  • C20:5
  • C22:4
  • C22:5
  • C22:6
  • Methyl arachidonate
  • Methyl eicosenoate
  • Methyl myristate