GC Analysis of FAMEs in Atlantic Salmon on SLB®-IL60
GC Analysis of FAMEs in Atlantic Salmon on SLB®-IL60
Conditions
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
oven 170 °C, 1 °C/min to 225 °C
carrier gas helium, 1.2 mL/min
sample extract of a farm raised Atlantic salmon, the antioxidant butylated hydroxytoluene (BHT) added prior to extraction
injection 1 µL100:1 split
liner 4 mm I.D., split/splitless type, wool packed single taper FocusLiner™ design
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Food and Beverages
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC