GC Analysis of FAMEs in B20 Biodiesel (Soy Source) on SLB®-IL111
GC Analysis of FAMEs in B20 Biodiesel (Soy Source) on SLB®-IL111
Conditions
column SLB-IL111, 30 m x 0.25 mm I.D., 0.20 µm (28927-U)
oven 50 C, 13 C/min to 270 C (5 min)
carrier gas helium, 40 cm/sec
sample B20 biodiesel (soy source) diluted 1:20 in n-hexane
injection 1 µL, 100:1 split
liner 4 mm I.D. FocusLiner inlet liner (no taper)
inj. temp. 250 C
detector FID, 270 C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Biofuels
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes