GC Analysis of FAMEs in Butter on Omegawax® (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of FAMEs in Butter on Omegawax® (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Omegawax, 10 m x 0.10 mm I.D., 0.10 µm (custom dimension)
oven 50 C, 90.0 C/min to 250 C
carrier gas hydrogen, 116.0 cm/sec, 400 kPa
sample butter FAMES, diluted 20:1 in hexane
injection 0.2 µL, 200:1 split
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Food and Beverages
Legal Information Omegawax is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes