Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of FAMEs in Canola Oil on SLB-IL111
GC Analysis of FAMEs in Canola Oil on SLB-IL111
Conditions
column SLB-IL111, 30 m x 0.25 mm I.D., 0.20 µm (28927-U)
oven (180 C)
carrier gas (helium, 25 cm/sec)
sample (Canola oil characterized reference oil (46961), methylated using BF3-methanol prior to analysis)
injection (1 µL, 50:1 split)
liner (4 mm I.D., split type, cup design)
inj. temp. (250 C)
detector (FID, 260 C)
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Other Notes Complementary products:
Syringes