GC Analysis of FAMEs in Cod Liver Oil on SUPELCOWAX® 10 (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of FAMEs in Cod Liver Oil on SUPELCOWAX® 10 (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SUPELCOWAX 10, 10 m x 0.10 mm I.D., 0.10 µm (25026-U)
oven 180 °C, 40 °C/min to 270 °C (0.5 min)
carrier gas hydrogen, 100 cm/sec constant
sample cod liver oil FAMEs in hexane
injection 0.2 µL in the split mode (200:1)
inj. temp. 280 °C
detector FID, 280 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Food and Beverages
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes