GC Analysis of FAMEs in Five Edible Oils (Canola, Lard, Palm, Soybean, and Sunflower Seed) on SLB®-IL111
GC Analysis of FAMEs in Five Edible Oils (Canola, Lard, Palm, Soybean, and Sunflower Seed) on SLB®-IL111
Conditions
column SLB-IL111, 30 m x 0.25 mm I.D., 0.20 µm (28927-U)
oven 180 °C
carrier gas helium, 25 cm/sec
sample Canola (46961), Lard (47115-U), Palm (46962), Soybean (47122), and Sunflower Seed (47123) characterized reference oils, methylated using BF3-methanol prior to analysis
injection 1 µL, 50:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes