GC Analysis of FAMEs in Palm Oil on SLB-IL111
GC Analysis of FAMEs in Palm Oil on SLB-IL111
Conditions
column SLB-IL111, 30 m x 0.25 mm I.D., 0.20 µm (28927-U)
oven 180 °C
carrier gas helium, 25 cm/sec
sample Characterized reference oils, methylated using BF3-methanolprior to analysis
injection 1 µL, 50:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Other Notes Complementary products:
Syringes