GC Analysis of FAMEs in Plasma on SUPELCOWAX® 10 (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of FAMEs in Plasma on SUPELCOWAX® 10 (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SUPELCOWAX 10, 10 m x 0.10 mm I.D., 0.10 µm (25026-U)
oven 220 °C, 60 °C/min to 280 °C (1 min)
carrier gas hydrogen, 120 cm/sec constant
sample plasma FAMEs in hexane
injection 0.5 µL, 30:1 split
inj. temp. 280 °C
detector FID, 280 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Clinical
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester
  • cis-8,11,14-Eicosatrienoic acid methyl ester
  • Methyl all-cis-5,8,11,14,17-eicosapentaenoate
  • Methyl all-cis-7,10,13,16,19-docosapentaenoate
  • Methyl arachidonate
  • Methyl heptadecanoate
  • Methyl linoleate
  • Methyl linolenate
  • Methyl myristate
  • Methyl oleate
  • Methyl palmitate
  • Methyl palmitoleate
  • Methyl pentadecanoate
  • Methyl stearate
  • Methyl cis-6,9,12-hexadecatrienoate