GC Analysis of FAMEs in Rapeseed Oil on SUPELCOWAX® 10
GC Analysis of FAMEs in Rapeseed Oil on SUPELCOWAX® 10
Conditions
column SUPELCOWAX 10, 30m x 0.53 mm I.D., 1.0 µm (25301-U)
oven 240 C
carrier gas helium, 5 mL/min (flow controlled)
sample Rapeseed Oil Reference mix (O7756-1AMP)
injection 0.1 muLdirect
inj. temp. 250 C
detector FID, 260 C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • C1:0
  • C16:0
  • C18:0
  • C18:1
  • C18:2
  • C18:3
  • C20:0
  • C20:1
  • C22:0
  • C22:1
  • C24:0
  • Methyl arachidate
  • Methyl behenate
  • Methyl eicosenoate
  • Methyl erucate
  • Methyl linolenate
  • Methyl myristate
  • Methyl oleate
  • Methyl palmitate
  • Methyl stearate