GC Analysis of FAMEs in Rapeseed Oil on SLB®-IL100
GC Analysis of FAMEs in Rapeseed Oil on SLB®-IL100
Conditions
column SLB-IL100, 30 m x 0.25 mm I.D., 0.20 µm (28884-U)
oven 180 °C
carrier gas helium, 30 cm/sec @ 180 °C
sample rapeseed oil FAME mix, 5 mg/mL total FAMEs in methylene chloride
injection 1 µL, 100:1 split
liner 4 mm I.D., split, cup
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes