GC Analysis of FAMEs in Sunflower Seed Oil on SLB®-IL111
GC Analysis of FAMEs in Sunflower Seed Oil on SLB®-IL111
Conditions
column SLB-IL111, 30 m x 0.25 mm I.D., 0.20 µm (28927-U7)
oven 180 °C
carrier gas helium, 25 cm/sec
sample Sunflower seed oil characterized reference oil (47123), methylated using BF3-methanol prior to analysis
injection 1 µL50:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes