GC Analysis of Fluorocarbons on a Packed Column (5% Fluorocol™ on 60/80 Carbopack™ B)
GC Analysis of Fluorocarbons on a Packed Column (5% Fluorocol™ on 60/80 °Carbopack™ B)
Conditions
column 10' x 1/8" SP™ alloy, general configuration, packed with 5% Fluorcol™ on 60/80 °Carbopack B
oven 50 °C
carrier gas nitrogen, 30 mL/min
injection 10 µLsynthetic mixture (approximately 20% each compound)
detector FID
Description
Analysis Note A packed GC column using Fluorcol stationary phase is also useful for fluorocarbons. The polarity of this column provides an alternate elution pattern compared to the Alumina chloride PLOT column chromatogram on the previous slide.
Categories Analytical Chromatography, GC Applications, Gases
Featured Industry Chemicals and Industrial Polymers
Legal Information Fluorcol is a trademark of Sigma-Aldrich Co. LLC, Carbopack is a trademark of Sigma-Aldrich Co. LLC, SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Dichlorofluoromethane
  • Freon 11
  • Freon 114
  • Freon 114a
  • Freon 12
  • Freon 21
  • Freon 22
  • Trichlorofluoromethane