GC Analysis of Fluoxetine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX® B-DA
GC Analysis of Fluoxetine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX® B-DA
Conditions
column Astec CHIRALDEX B-DA, 10 m x 0.25 mm I.D., 0.12 µm (72021AST)
oven 175 °C
carrier gas helium, 15 psi
sample peaks 1 & 2: fluoxetine enantiomers (N-TFA derivatives)
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Chiral Applications, Clinical, Forensic & Toxicology, Enantiomers, Pharmaceuticals, Therapeutic Drug Monitoring
Featured Industry Clinical
Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes