GC Analysis of Formalin Solution on Carboxen®-1006 PLOT
GC Analysis of Formalin Solution on Carboxen®-1006 PLOT
Conditions
column Carboxen-1006 PLOT
oven 200 C
carrier gas 3.0 ml/min
sample Formalin solution
injection 0.5 µl
detector TCD
Description
Categories Analytical Chromatography, GC Applications, Solvents, Aldehydes, Alcohols
Featured Industry Chemicals and Industrial Polymers
Legal Information Carboxen is a registered trademark of Sigma-Aldrich Co. LLC