GC Analysis of Glycerides in Palm Oil on MET-Biodiesel
GC Analysis of Glycerides in Palm Oil on MET-Biodiesel
Conditions
column MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µm with integrated 2 m x 0.53 mm I.D. guard (28668-U)
oven 200 °C, 20 °C/min to 350 °C (15 min)
carrier gas helium, 10 mL/min
sample palm oil extract (subsequent derivatization with TMS)
injection 1 µL, cool on-column
detector FID, 380 °C
Description
Categories Analytical Chromatography, GC Applications, Glycerides
Featured Industry Agriculture
Food and Beverages
Other Notes Complementary products:
Syringes