GC Analysis of Glycerin Impurity in B100 Biodiesel (with I.S.) on MET-Biodiesel
GC Analysis of Glycerin Impurity in B100 Biodiesel (with I.S.) on MET-Biodiesel
Conditions
column MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µm with integrated 2 m x 0.53 mm I.D. guard (28668-U)
oven 50 °C (1 min), 15 °C/min to 180 °C, 7 °C/min. to 230 °C, 30 °C/min to 380 °C (10 min)
carrier gas helium, 3.0 mL/min
sample B100 Biodiesel plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA then diluted in heptane
injection 1 µL, cold on-column
detector FID, 380 °C
Description
Categories Analytical Chromatography, GC Applications, Glycerides
Featured Industry Biofuels
Other Notes Complementary products:
Syringes