Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Glycols and Diols on SPB®-1000
GC Analysis of Glycols and Diols on SPB®-1000
Conditions
column SPB-1000, 30 m x 0.53 mm I.D., 0.50 µm (25445)
oven 50 C (2 min), 8 C/min to 200 C (2 min)
carrier gas helium, 37 cm/sec, constant
sample each analyte 100 ppm in methanol
injection 1 µL, pulsed (10 psi until 0.10 min) splitless (0.75 min)
liner 2 mm I.D., straight
inj. temp. 250 C
detector FID, 300 C
Description
Categories Analytical Chromatography, GC Applications, Diols, Glycols
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,3-Propanediol
  • 1,4-Butanediol
  • 2-(2-Butoxyethoxy)ethanol
  • 2-(2-Ethoxyethoxy)ethanol
  • 2,3-Butanediol
  • 2-Butoxyethanol
  • 2-Ethoxyethanol
  • 2-Methoxyethanol
  • Dipropylene glycol
  • Ethylene glycol
  • Propylene glycol