Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Glycols and Diols on Equity®-1
GC Analysis of Glycols and Diols on Equity®-1
Conditions
column Equity-1, 30 m x 0.53 mm I.D., 0.50 µm (28068-U)
oven 50 C (2 min), 8 C/min to 200 C (2 min)
carrier gas helium, 37 cm/sec, constant
sample each analyte at 100 ppm in methanol
injection 1 µL, pulsed (10 psi until 0.10 min) splitless (0.75 min)
liner 2 mm I.D., straight
inj. temp. 250 C
detector FID, 300 C
Description
Categories Analytical Chromatography, GC Applications, Diols, Glycols
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes