GC Analysis of α-Hexachlorocyclohexane (α-HCH) Enantiomers on Astec® CHIRALDEX® G-PN
GC Analysis of α-Hexachlorocyclohexane (α-HCH) Enantiomers on Astec® CHIRALDEX® G-PN
Conditions
column Astec CHIRALDEX G-PN, 30 m x 0.25 mm I.D., 0.12 µm (74033AST)
oven 170 °C
carrier gas helium, 30 psi
sample 2 mg/mL in ethanolpeaks 1 & 2: alpha-HCH (alpha-hexachlorocyclohexane) enantiomers
injection 0.2 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Pesticides
Featured Industry Agriculture
Chemicals and Industrial Polymers
Environmental
Food and Beverages
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes