Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of α-Hexachlorocyclohexane (α-HCH) Enantiomers on Astec® CHIRALDEX® G-DM
GC Analysis of α-Hexachlorocyclohexane (α-HCH) Enantiomers on Astec® CHIRALDEX® G-DM
Conditions
column Astec CHIRALDEX B-DM, 30 m x 0.25 mm I.D., 0.12 µm (77023AST)
oven 170 C
carrier gas helium, 30 psi
sample 2 mg/mL in ethanolpeaks 1 & 2: alpha-HCH (alpha-hexachlorocyclohexane) enantiomers
injection 0.2 µL, 80:1 split
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Pesticides
Featured Industry Agriculture
Chemicals and Industrial Polymers
Environmental
Food and Beverages
Legal Information CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes