GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH, Isothermal
GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH, Isothermal
Conditions
column Petrocol DH, 100 m x 0.25 mm I.D., 0.50 µm (24160-U)
oven 60 °C
carrier gas helium, 19-21 cm/sec
injection 0.1 µL, 100:1 split
inj. temp. 250 °C
detector FID, 250 ºC
Description
Analysis Note Detailed hydrocarbon analysis (DHA) is performed on long non-polar columns. Elution occurs in order of lowest boiling point to highest. The chromatogram shown here is of a gasoline sample using a 100 m Petrocol DH column. The long column length and 60 ºC isothermal oven temperature allow the separation of many components in this complex sample. Compounds identified range from propane (peak 1) to 1-methylnaphthalene (peak 96). Retention index libraries exist for this column, adding to its usefulness.
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Petroleum
Legal Information Petrocol is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,3,4-Tetramethylbenzene
  • 1,2,3,5-Tetramethylbenzene
  • 1,2,3-Trimethylbenzene
  • 1,2,4,5-Tetramethylbenzene
  • 1,2-Dimethyl-4-ethylbenzene
  • 1,3,5-Trimethylbenzene
  • 1,3-Diethylbenzene
  • 1,3-Dimethyl-4-ethylbenzene
  • 1,3-Dimethyl-5-ethylbenzene
  • 1,4-Diethylbenzene
  • 1,4-Dimethyl-2-ethylbenzene
  • 1-Methyl-2-ethylbenzene
  • 1-Methyl-3-ethylbenzene
  • 1-Methyl-4-ethylbenzene
  • 1-Methylnaphthalene
  • 1-Nonene
  • 1-Pentene
  • 1-Undecene
  • 2,2,4-Trimethylpentane
  • 2,2-Dimethylbutane
  • 2,2-Dimethylheptane
  • 2,3,4-Trimethylpentane
  • 2,3-Dimethylbutane
  • 2,3-Dimethylhexane
  • 2,3-Dimethylpentane
  • 2,4,5-Trimethylheptane
  • 2,4-Dimethylheptanen
  • 2,4-Dimethylhexane
  • 2,4-Dimethylpentane
  • 2,5-Dimethylhexane
  • 2,6-Dimethylheptanen
  • 2-Methyl-1-butene
  • 2-Methyl-1-heptene
  • 2-Methyl-1-hexene
  • 2-Methyl-2-butene
  • 2-Methylheptane
  • 2-Methylhexene
  • 2-Methyllindane
  • 2-Methylnaphthalene
  • 2-Methyloctane
  • 2-Methylpentane
  • 3,3,4-Trimethylheptane
  • 3,3,5-Trimethylheptane
  • 3,4-Dimethyl-1-hexene
  • 3,4-Dimethyl-1-hexenen
  • 3,4-Dimethylhexane
  • 3,4-Dimethyl-trans-hexene
  • 3-Ethyl-3-methylpentane
  • 3-Ethylcyclopentane
  • 3-Ethylheptane
  • 3-Methyl-1-Butene
  • 3-Methyl-2-pentene
  • 3-Methylcyclopentene
  • 3-Methylheptane
  • 3-Methylhexane
  • 3-Methylnonane
  • 3-Methyloctane
  • 3-Methylpentane
  • 3-Methyl-trans-2-pentene
  • 4-Methylheptane
  • 4-Methyloctane
  • 5-Methyl-1-hexene
  • Benzene
  • Butane
  • cis-2-Butene
  • cis-2-Hexene
  • cis-2-Pentene
  • Cyclohexane
  • Cyclopentene
  • Ethylbenzene
  • Ethylcyclohexane
  • Hexane
  • Indan
  • Isobutane
  • Isobutylbenzene
  • Isobutylene
  • Isopentane
  • Isopropylbenzene (cumene)
  • Methylcyclopentane
  • m-Xylene
  • Naphthalene
  • n-Butylbenzene
  • n-Decane
  • n-Heptane
  • n-Nonane
  • n-Octane
  • n-Propylbenzene
  • o-Xylene
  • Pentane
  • Propane
  • p-Xylene
  • sec-Butylbenzene
  • Toluene
  • trans-2-Pentene