GC Analysis of Hydrocarbons in Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Hydrocarbons in Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Equity-1, 15 m x 0.10 mm I.D., 0.10 µm (28039-U)
oven 40 C (1 min), 45 C/min to 150 C (2 min)
carrier gas hydrogen, 45 cm/sec constant
sample Unleaded gasoline (refinery standard), neat
injection 0.1 muL300:1 split
liner 2 mm I.D., straight
inj. temp. 175 C
detector FID, 175 C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Petroleum
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1,2,3,5-Tetramethylbenzene
  • 1,2,3-Trimethylbenzene
  • 1,2,4,5-Tetramethylbenzene
  • 1,2,4-Trimethylbenzene
  • 1,2-Dimethyl-4-ethylbenzene
  • 1,3,5-Trimethylbenzene
  • 1,3-Diethylbenzene
  • 1,3-Dimethyl-4-ethylbenzene
  • 1,4-Dimethyl-2-ethylbenzene
  • 1-Methyl-2-ethylbenzene
  • 1-Methyl-3-ethylbenzene
  • 1-Methyl-4-ethylbenzene
  • 1-Methylnaphthalene
  • 2,2,4-Trimethylpentane
  • 2,2-Dimethylbutane
  • 2,3,4-Trimethylpentane
  • 2,3-Dimethylbutane
  • 2,3-Dimethylhexane
  • 2,3-Dimethylpentane
  • 2,4-Dimethylhexane
  • 2,4-Dimethylpentane
  • 2,5-Dimethylhexane
  • 2.3-Dimethylpentane
  • 2-Methylbutane
  • 2-Methylheptane
  • 2-Methylhexane
  • 2-Methylnaphthalene
  • 2-Methylpentane
  • 2-Methylpropane
  • 3,3,4-Trimethylheptane
  • 3-Methylheptane
  • 3-Methylhexane
  • 3-Methylpentane
  • Benzene
  • Butane
  • Dimethylnaphthalenes
  • Durene
  • Ethylbenzene
  • Heptane
  • Hexane
  • iso-Butylbenzene
  • Isooctane
  • Isopropylbenzene (cumene)
  • lndane
  • lsobutane
  • lsopentane
  • m,p-Xylene
  • Mesitylene
  • m-Xylene
  • Naphthalene
  • N-Butylbenzene
  • Nonane
  • Octane
  • o-Xylene
  • Pentane
  • Propylbenzene
  • p-Xylene
  • sec-Butylbenzene
  • Toluene